B-[6-(3-Methoxypropoxy)-3-pyridinyl]boronic acid ≥95%

Supplier: AOBChem USA

Warning

180296-1G 180296-500MG 180296-5G
77466-008EA 597.4 USD
77466-008 77478-752 77488-974
B-[6-(3-Methoxypropoxy)-3-pyridinyl]boronic acid ≥95%
B-[6-(3-Methoxypropoxy)-3-pyridinyl]boronic acid

Precaución: For research use only

Formula: C₉H₁₄BNO₄
MW: 211.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28335081
CAS Number: 1334477-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR