4-(Hydroxymethyl)-2,6-dimethylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18010-1G 18010-500MG 18010-5G 18010-25G
77466-004EA 106.09 USD
77466-004 77478-748 77488-968 77499-458
4-(Hydroxymethyl)-2,6-dimethylphenylboronic acid ≥97%
4-(Hydroxymethyl)-2,6-dimethylphenylboronic acid

Precaución: For research use only

Formula: C₉H₁₃BO₃
MW: 180.009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713089
CAS Number: 2121513-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR