6-Chloro-2-(trifluoromethyl)nicotinonitrile ≥96%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-2-(trifluoromethyl)-3-pyridinecarbonitrile

179994-1G 179994-500MG 179994-5G 179994-25G
77466-000EA 170.98 USD
77466-000 77478-744 77488-964 77499-452
6-Chloro-2-(trifluoromethyl)nicotinonitrile ≥96%
6-Chloro-2-(trifluoromethyl)nicotinonitrile

Precaución: For research use only

Formula: C₇H₂ClF₃N₂
MW: 206.552 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848371
CAS Number: 1245913-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR