1,3-Bis(1,1-dimethylethyl) 2-(5-borono-3-methoxy-2-pyridinyl)imidodicarbonate ≥95%

Supplier: AOBChem USA

Warning

17954-1G 17954-500MG 17954-5G 17954-25G
77465-996EA 226.6 USD
77465-996 77478-740 77488-960 77499-448
1,3-Bis(1,1-dimethylethyl) 2-(5-borono-3-methoxy-2-pyridinyl)imidodicarbonate ≥95%
1,3-Bis(1,1-dimethylethyl) 2-(5-borono-3-methoxy-2-pyridinyl)imidodicarbonate

Precaución: For research use only

Formula: C₁₆H₂₅BN₂O₇
MW: 368.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755719
CAS Number: 1111637-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR