2-Fluoro-4,5-dimethylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

17934-1G 17934-500MG 17934-5G
77465-992EA 801.34 USD
77465-992 77478-736 77488-956
2-Fluoro-4,5-dimethylphenylboronic acid ≥95%
2-Fluoro-4,5-dimethylphenylboronic acid

Precaución: For research use only

Formula: C₈H₁₀BFO₂
MW: 167.973 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22189552
CAS Number: 1125394-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR