2,6-Dichloro-3-hydroxyisonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

17876-1G 17876-500MG 17876-5G 17876-25G
77465-988EA 385.22 USD
77465-988 77478-732 77488-952 77499-440
2,6-Dichloro-3-hydroxyisonicotinaldehyde ≥97%
2,6-Dichloro-3-hydroxyisonicotinaldehyde

Precaución: For research use only

Formula: C₆H₃Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD06410675
CAS Number: 185423-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR