1,2-Dimethyl-3-(methylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

178737-1G 178737-500MG 178737-5G
77465-984EA 648.9 USD
77465-984 77478-728 77488-948
1,2-Dimethyl-3-(methylthio)benzene ≥95%
1,2-Dimethyl-3-(methylthio)benzene

Precaución: For research use only

Formula: C₉H₁₂S
MW: 152.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617794
CAS Number: 66794-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR