6-Bromo-3-fluoropicolinic acid ≥97%

Supplier: AOBChem USA

Warning

177984-1G 177984-500MG 177984-5G 177984-25G
77465-980EA 93.73 USD
77465-980 77478-724 77488-944 77499-432
6-Bromo-3-fluoropicolinic acid ≥97%
6-Bromo-3-fluoropicolinic acid

Precaución: For research use only

Formula: C₆H₃BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD13185805
CAS Number: 1052714-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR