1-Bromo-3-isobutylbenzene ≥95%

Supplier: AOBChem USA

Warning

177520-1G 177520-5G 177520-25G
77465-976EA 67.98 USD
77465-976 77488-940 77499-430
1-Bromo-3-isobutylbenzene ≥95%
1-Bromo-3-isobutylbenzene

Precaución: For research use only

Formula: C₁₀H₁₃Br
Storage Temperature: Ambient
MDL Number: MFCD26405543
CAS Number: 139155-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR