1-(2-Bromo-4-ethylphenyl)ethanol ≥96%

Supplier: AOBChem USA

Warning

177512-1G 177512-500MG 177512-5G 177512-25G
77465-972EA 415.09 USD
77465-972 77478-718 77488-936 77499-426
1-(2-Bromo-4-ethylphenyl)ethanol ≥96%
1-(2-Bromo-4-ethylphenyl)ethanol

Precaución: For research use only

Formula: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD31707019
CAS Number: 2969219-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR