2,3-Dichloro-1-fluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

177495-1G 177495-500MG 177495-5G 177495-25G
77465-968EA 147.29 USD
77465-968 77478-714 77488-932 77499-422
2,3-Dichloro-1-fluoro-4-methoxybenzene ≥97%
2,3-Dichloro-1-fluoro-4-methoxybenzene

Precaución: For research use only

Formula: C₇H₅Cl₂FO
MW: 195.018 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19690559
CAS Number: 1378832-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR