3-Bromo-5-(trifluoromethoxy)anisole ≥95%

Supplier: AOBChem USA

Warning

17702-1G 17702-500MG 17702-5G 17702-25G
77465-964EA 106.09 USD
77465-964 77478-710 77488-928 77499-418
3-Bromo-5-(trifluoromethoxy)anisole ≥95%
3-Bromo-5-(trifluoromethoxy)anisole

Precaución: For research use only

Formula: C₈H₆BrF₃O₂
MW: 271.031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19981548
CAS Number: 1330750-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR