(3-Fluoro-4-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

176976-1G 176976-500MG 176976-5G
77465-960EA 648.9 USD
77465-960 77478-706 77488-924
(3-Fluoro-4-methylphenyl)(methyl)sulfane ≥95%
(3-Fluoro-4-methylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₈H₉FS
Storage Temperature: Ambient
MDL Number: MFCD11617748
CAS Number: 142994-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR