3'-bromo-2'-fluoro-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol ≥97%

Supplier: AOBChem USA

Warning

176901-1G 176901-500MG 176901-5G 176901-25G
77465-956EA 340.93 USD
77465-956 77478-702 77488-920 77499-412
3'-bromo-2'-fluoro-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol ≥97%
3'-bromo-2'-fluoro-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol

Precaución: For research use only

Formula: C₁₂H₁₂BrFO
MW: 271.1254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34855059
CAS Number: 1694006-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR