4'-bromo-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol ≥95%

Supplier: AOBChem USA

Warning

176894-1G 176894-500MG 176894-5G 176894-25G
77465-952EA 255.44 USD
77465-952 77478-698 77488-916 77499-408
4'-bromo-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol ≥95%
4'-bromo-1,2,3,4-tetrahydro-[1,1'-biphenyl]-1-ol

Precaución: For research use only

Formula: C₁₂H₁₃BrO
MW: 253.1350 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34855056
CAS Number: 2002017-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR