(2-Fluoro-5-methylphenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

176882-1G 176882-500MG 176882-5G 176882-25G
77465-948EA 415.09 USD
77465-948 77478-694 77488-912 77499-404
(2-Fluoro-5-methylphenyl)(isopropyl)sulfane ≥95%
(2-Fluoro-5-methylphenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₃FS
MW: 184.2736 g/mol
Storage Temperature: Ambient
CAS Number: 3002464-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR