5-Bromo-2-(trifluoromethyl)-4-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-trifluoromethylpyridine-4-boronic acid

17616-1G 17616-5G 17616-25G
77465-944EA 65.92 USD
77465-944 77488-908 77499-400
5-Bromo-2-(trifluoromethyl)-4-pyridineboronic acid ≥97%
5-Bromo-2-(trifluoromethyl)-4-pyridineboronic acid

Precaución: For research use only

Formula: C₆H₄BBrF₃NO₂
MW: 269.81 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11504859
CAS Number: 1072951-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR