1-(3-Bromo-5-methyl-2-thienyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

175792-1G 175792-500MG 175792-5G
77465-936EA 466.59 USD
77465-936 77478-684 77488-900
1-(3-Bromo-5-methyl-2-thienyl)-2,2,2-trifluoroethanone ≥95%
1-(3-Bromo-5-methyl-2-thienyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₇H₄BrF₃OS
MW: 273.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107310
CAS Number: 1504916-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR