2-Fluoro-6-(methylsulfonyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

175740-1G 175740-500MG 175740-5G 175740-25G
77465-932EA 277.07 USD
77465-932 77478-680 77488-896 77499-392
2-Fluoro-6-(methylsulfonyl)pyridine ≥95%

Precaución: For research use only

Formula: C₆H₆FNO₂S
Storage Temperature: Ambient
MDL Number: MFCD19689741
CAS Number: 1207609-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR