4-(3-Bromophenoxy)aniline ≥97%

Supplier: AOBChem USA

Warning

175234-1G 175234-500MG 175234-5G 175234-25G
77465-928EA 204.97 USD
77465-928 77478-676 77488-890 77499-388
4-(3-Bromophenoxy)aniline ≥97%
4-(3-Bromophenoxy)aniline

Precaución: For research use only

Formula: C₁₂H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD02663382
CAS Number: 57688-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR