1-(4-Chlorophenyl)-2-methylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

175225-1G 175225-500MG 175225-5G 175225-25G
77465-920EA 169.95 USD
77465-920 77478-668 77488-882 77499-380
1-(4-Chlorophenyl)-2-methylbutan-1-ol ≥97%
1-(4-Chlorophenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅ClO
MW: 198.6892 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12786867
CAS Number: 14248-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR