1-(3-Bromophenyl)-2-methylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

175221-1G 175221-500MG 175221-5G 175221-25G
77465-916EA 415.09 USD
77465-916 77478-664 77488-878 77499-376
1-(3-Bromophenyl)-2-methylbutan-1-ol ≥97%
1-(3-Bromophenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅BrO
MW: 243.1402 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21928925
CAS Number: 1517266-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR