1-(5-Bromo-2-fluorophenyl)-2-methylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

175215-1G 175215-500MG 175215-5G 175215-25G
77465-912EA 415.09 USD
77465-912 77478-660 77488-874 77499-372
1-(5-Bromo-2-fluorophenyl)-2-methylbutan-1-ol ≥97%
1-(5-Bromo-2-fluorophenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄BrFO
MW: 261.1306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21928930
CAS Number: 1515938-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR