1-(2-Fluoro-5-methoxyphenyl)-2-methylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

175211-1G 175211-500MG 175211-5G 175211-25G
77465-908EA 415.09 USD
77465-908 77478-656 77488-870 77499-368
1-(2-Fluoro-5-methoxyphenyl)-2-methylbutan-1-ol ≥97%
1-(2-Fluoro-5-methoxyphenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₇FO₂
MW: 212.2605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34854568
CAS Number: 2969432-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR