4-(2-Fluoro-5-methoxyphenyl)tetrahydro-2H-thiopyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

175207-1G 175207-500MG 175207-5G 175207-25G
77465-904EA 415.09 USD
77465-904 77478-652 77488-866 77499-364
4-(2-Fluoro-5-methoxyphenyl)tetrahydro-2H-thiopyran-4-ol ≥97%
4-(2-Fluoro-5-methoxyphenyl)tetrahydro-2H-thiopyran-4-ol

Precaución: For research use only

Formula: C₁₂H₁₅FO₂S
MW: 242.3097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34854566
CAS Number: 2969329-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR