1-(2-Fluoro-5-methoxyphenyl)-3-methylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

175202-1G 175202-500MG 175202-5G
77465-900EA 447.02 USD
77465-900 77478-648 77488-862
1-(2-Fluoro-5-methoxyphenyl)-3-methylbutan-1-ol ≥97%
1-(2-Fluoro-5-methoxyphenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₇FO₂
MW: 212.2605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34854563
CAS Number: 1557590-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR