1-(3-Bromo-2,6-difluorophenyl)-3-methylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

175193-1G 175193-500MG 175193-5G
77465-896EA 447.02 USD
77465-896 77478-644 77488-858
1-(3-Bromo-2,6-difluorophenyl)-3-methylbutan-1-ol ≥97%
1-(3-Bromo-2,6-difluorophenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₃BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34854561
CAS Number: 1700519-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR