4-(3-Fluoro-5-methoxyphenyl)tetrahydro-2H-thiopyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

175171-1G 175171-500MG 175171-5G 175171-25G
77465-892EA 312.09 USD
77465-892 77478-640 77488-854 77499-358
4-(3-Fluoro-5-methoxyphenyl)tetrahydro-2H-thiopyran-4-ol ≥95%
4-(3-Fluoro-5-methoxyphenyl)tetrahydro-2H-thiopyran-4-ol

Precaución: For research use only

Formula: C₁₂H₁₅FO₂S
MW: 242.3097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34854558
CAS Number: 2007736-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR