4-(4-Chlorophenyl)tetrahydro-2H-thiopyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

175163-1G 175163-500MG 175163-5G 175163-25G
77465-888EA 328.57 USD
77465-888 77478-636 77488-850 77499-354
4-(4-Chlorophenyl)tetrahydro-2H-thiopyran-4-ol ≥97%
4-(4-Chlorophenyl)tetrahydro-2H-thiopyran-4-ol

Precaución: For research use only

Formula: C₁₁H₁₃ClOS
MW: 228.7383 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31693986
CAS Number: 853886-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR