4-(3-Bromo-2-fluorophenyl)tetrahydro-2H-thiopyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

174763-1G 174763-500MG 174763-5G 174763-25G
77465-884EA 340.93 USD
77465-884 77478-632 77488-846 77499-350
4-(3-Bromo-2-fluorophenyl)tetrahydro-2H-thiopyran-4-ol ≥97%
4-(3-Bromo-2-fluorophenyl)tetrahydro-2H-thiopyran-4-ol

Precaución: For research use only

Formula: C₁₁H₁₂BrFOS
MW: 291.1797 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34854355
CAS Number: 2008375-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR