3-Bromo-5-methyl-2-thiophenecarboxylic acid ≥95%

Supplier: AOBChem USA

Warning

174645-1G 174645-500MG 174645-5G 174645-25G
77465-876EA 260.59 USD
77465-876 77478-624 77488-838 77499-346
3-Bromo-5-methyl-2-thiophenecarboxylic acid ≥95%
3-Bromo-5-methyl-2-thiophenecarboxylic acid

Precaución: For research use only

Formula: C₆H₅BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD01859872
CAS Number: 61285-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR