2-Fluoro-4-(trifluoromethoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-4-(trifluoromethoxy)benzoic acid

174352-1G 174352-5G 174352-25G
77465-872EA 60.77 USD
77465-872 77488-834 77499-340
2-Fluoro-4-(trifluoromethoxy)benzoic acid ≥95%
2-Fluoro-4-(trifluoromethoxy)benzoic acid

Precaución: For research use only

Formula: C₈H₄F₄O₃
MW: 224.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652439
CAS Number: 1073477-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR