2-Bromo-5-fluoro-α,α-dimethyl-4-pyridinemethanol ≥95%
Supplier: AOBChem USA
|
Warning
|
173675-1G
173675-500MG
173675-5G
173675-25G
77465-868EA
415.09
USD
77465-868
77478-618
77488-830
77499-336
2-Bromo-5-fluoro-α,α-dimethyl-4-pyridinemethanol ≥95%
2-Bromo-5-fluoro-α,α-dimethyl-4-pyridinemethanol
Precaución: For research use only
Formula:
C₈H₉BrFNO MW: 234.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23163846 CAS Number: 1387560-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...