1-[4-(Ethylthio)phenyl]-1-propanone ≥95%

Supplier: AOBChem USA

Warning

173195-1G 173195-500MG 173195-5G
77465-860EA 576.8 USD
77465-860 77478-612 77488-822
1-[4-(Ethylthio)phenyl]-1-propanone ≥95%
1-[4-(Ethylthio)phenyl]-1-propanone

Precaución: For research use only

Formula: C₁₁H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD06590180
CAS Number: 69567-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR