1-Butyl-4-(methylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

173180-1G 173180-500MG 173180-5G
77465-856EA 648.9 USD
77465-856 77478-608 77488-818
1-Butyl-4-(methylthio)benzene ≥95%
1-Butyl-4-(methylthio)benzene

Precaución: For research use only

Formula: C₁₁H₁₆S
MW: 180.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617762
CAS Number: 252555-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR