4'-ethyl-[1,1'-biphenyl]-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

172864-1G 172864-500MG 172864-5G 172864-25G
77465-848EA 141.11 USD
77465-848 77478-602 77488-806 77499-314
4'-ethyl-[1,1'-biphenyl]-4-carbaldehyde ≥95%
4'-ethyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: For research use only

Formula: C₁₅H₁₄O
MW: 210.2710 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017714
CAS Number: 101002-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR