Ethanone, 1-(2,4-difluoro-3-hydroxyphenyl)- ≥97%
Supplier: AOBChem USA
|
Warning
|
17213-1G
17213-500MG
17213-5G
17213-25G
77465-844EA
426.42
USD
77465-844
77478-598
77488-802
77499-306
Ethanone, 1-(2,4-difluoro-3-hydroxyphenyl)- ≥97%
Ethanone, 1-(2,4-difluoro-3-hydroxyphenyl)-
Precaución: For research use only
Formula:
C₈H₆F₂O₂ MW: 172.129 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16619378 CAS Number: 951163-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...