1,2,4-Trichloro-3-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

171457-1G 171457-500MG 171457-5G
77465-840EA 434.66 USD
77465-840 77478-594 77488-796
1,2,4-Trichloro-3-fluorobenzene ≥95%
1,2,4-Trichloro-3-fluorobenzene

Precaución: For research use only

Formula: C₆H₂Cl₃F
Storage Temperature: Ambient
MDL Number: MFCD18400143
CAS Number: 36556-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR