3-(1,1-Dimethylethyl)-2-ethoxy-5-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

170571-1G 170571-500MG 170571-5G
77465-832EA 736.45 USD
77465-832 77478-586 77488-786
3-(1,1-Dimethylethyl)-2-ethoxy-5-methylbenzaldehyde ≥95%
3-(1,1-Dimethylethyl)-2-ethoxy-5-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₄H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD07433280
CAS Number: 944664-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR