1-(3-Bromo-2,6-difluorophenyl)-2-methylbutan-1-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
170121-1G
170121-500MG
170121-5G
77465-828EA
447.02
USD
77465-828
77478-582
77488-780
1-(3-Bromo-2,6-difluorophenyl)-2-methylbutan-1-ol ≥95%
1-(3-Bromo-2,6-difluorophenyl)-2-methylbutan-1-ol
Precaución: For research use only
Formula:
C₁₁H₁₃BrF₂O Storage Temperature: Ambient |
MDL Number:
MFCD34853038 CAS Number: 1696677-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...