1-(2-Bromo-6-fluorophenyl)-2-methylbutan-1-ol ≥96%

Supplier: AOBChem USA

Warning

170117-1G 170117-500MG 170117-5G 170117-25G
77465-824EA 397.58 USD
77465-824 77478-578 77488-776 77499-288
1-(2-Bromo-6-fluorophenyl)-2-methylbutan-1-ol ≥96%
1-(2-Bromo-6-fluorophenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄BrFO
MW: 261.1306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34853037
CAS Number: 1598067-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR