1-(2-Fluorophenyl)butan-1-ol ≥96%

Supplier: AOBChem USA

Warning

170100-1G 170100-500MG 170100-5G
77465-820EA 471.74 USD
77465-820 77478-574 77488-772
1-(2-Fluorophenyl)butan-1-ol ≥96%
1-(2-Fluorophenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₃FO
MW: 168.2080 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16301262
CAS Number: 703-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR