1-(2-Fluorophenyl)-2-methylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

170090-1G 170090-500MG 170090-5G 170090-25G
77465-816EA 415.09 USD
77465-816 77478-570 77488-768 77499-282
1-(2-Fluorophenyl)-2-methylbutan-1-ol ≥97%
1-(2-Fluorophenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅FO
MW: 182.2346 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18871631
CAS Number: 1343248-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR