2-Methyl-1-phenylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

170086-1G 170086-500MG 170086-5G 170086-25G
77465-812EA 297.67 USD
77465-812 77478-566 77488-764 77499-278
2-Methyl-1-phenylbutan-1-ol ≥97%
2-Methyl-1-phenylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD00060860
CAS Number: 3968-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR