1-(4-Fluorophenyl)-3-methylbutan-1-ol ≥97%

Supplier: AOBChem USA

Warning

170082-1G 170082-500MG 170082-5G 170082-25G
77465-808EA 415.09 USD
77465-808 77478-562 77488-760 77499-274
1-(4-Fluorophenyl)-3-methylbutan-1-ol ≥97%
1-(4-Fluorophenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅FO
MW: 182.2346 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11934960
CAS Number: 92146-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR