1-(3-Methoxyphenyl)-3-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

170078-1G 170078-500MG 170078-5G 170078-25G
77465-804EA 297.67 USD
77465-804 77478-558 77488-756 77499-270
1-(3-Methoxyphenyl)-3-methylbutan-1-ol ≥95%
1-(3-Methoxyphenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD12153460
CAS Number: 78682-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR