1-(4-Bromophenyl)-3-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

170072-1G 170072-500MG 170072-5G 170072-25G
77465-800EA 297.67 USD
77465-800 77478-554 77488-752 77499-266
1-(4-Bromophenyl)-3-methylbutan-1-ol ≥95%
1-(4-Bromophenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅BrO
MW: 243.1402 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11920270
CAS Number: 100757-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR