1-(6-Bromo-5-methylpyridin-3-yl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

170057-1G 170057-500MG 170057-5G 170057-25G
77465-792EA 415.09 USD
77465-792 77478-546 77488-744 77499-258
1-(6-Bromo-5-methylpyridin-3-yl)cyclohexanol ≥97%
1-(6-Bromo-5-methylpyridin-3-yl)cyclohexanol

Precaución: For research use only

Formula: C₁₂H₁₆BrNO
MW: 270.1655 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34853029
CAS Number: 2921909-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR