(3-Chlorophenyl)(isopropyl)sulfane ≥96%

Supplier: AOBChem USA

Warning

170051-1G 170051-500MG 170051-5G
77465-788EA 450.11 USD
77465-788 77478-542 77488-740
(3-Chlorophenyl)(isopropyl)sulfane ≥96%
(3-Chlorophenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₉H₁₁ClS
Storage Temperature: Ambient
MDL Number: MFCD26792618
CAS Number: 55698-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR