5-Bromo-4-chloro-2-iodoanisole ≥96%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2-chloro-4-iodo-5-methoxybenzene

170001-1G 170001-500MG 170001-5G 170001-25G
77465-784EA 303.85 USD
77465-784 77478-538 77488-736 77499-254
5-Bromo-4-chloro-2-iodoanisole ≥96%
5-Bromo-4-chloro-2-iodoanisole

Precaución: For research use only

Formula: C₇H₅BrClIO
Storage Temperature: Ambient
MDL Number: MFCD11845928
CAS Number: 1160574-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR